Principaux documents
Z740021
BRAND® filter tips, racked, TipRack
volume 2-20 μL, non-sterile
About This Item
Code UNSPSC :
41121601
Nomenclature NACRES :
NB.01
Matériaux
colorless
colorless polypropylene tip and filter element (polyethylene)
Stérilité
non-sterile
Caractéristiques
RNase and DNase free
endotoxin free
filter
Conditionnement
pack of 960 ea (10 racks of 96)
Fabricant/nom de marque
BRAND 732608
Volume maximum
20 μL
Volume
2-20 μL
Description générale
Pipette tips and filter tips are manufactured by BRAND in a cleanroom under the most modern production conditions, and are automatically rack packed and packaged to ensure that the tips are of consistently high quality.
Non-self-sealing filter tips from BRAND have a PE filter that is free from chemical additives. Permeability is controlled by the combination of pore size and filter length, so that no aerosols can reach the pipette shaft. These filters function with consistent reliability. On the other hand, liquids can pass very slowly should they accidentally contact the filter.
Since the filter does not swell as in the case of self-sealing filter tips, the sample can be recovered from the filter by simply actuating the pipette′s blow-out function, or by centrifugation if necessary. This is clearly an important advantage of non-self-sealing filters, especially when working with valuable samples. As an added advantage, the absence of filter additives protects samples from this source of contamination.
NEW! TipRack, non-sterile and sterile, BIO-CERT®
Space-saving refill unit for TipBox. Protected in an environmentally compatible packaging of recyclable PET. Compared to the filled TipBoxes, the amount of waste with the new refill units is reduced by 20%. All pipette tips and filter tips up to 1000 μL are free of DNA, RNases, endotoxins and ATP. TipRacks in BIO-CERT® quality are sterile according to ISO 11 137 and the AAMI guidelines, a SAL of 10-6 is obtained.
These racks are supplied with a transfer aid that enables simple, contamination-free transfer into a previously sterilized TipBox. All tip-trays are printed on one side with information on the contents.
Autres remarques
BR732608 (India)
Informations légales
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG