Principaux documents
Z334103
BRAND® cryovial racks, PP
yellow
About This Item
Code UNSPSC :
41122806
Nomenclature NACRES :
NB.35
Matériaux
polypropylene
yellow
Caractéristiques
positions 6 × 14
Fabricant/nom de marque
BRAND 4341003
BRAND
Technique(s)
cryopreservation: suitable
L × l × H
265 mm × 126 mm × 38 mm
Diamètre
13 mm
Description générale
Racks for cryo vials are stackable, with alphanumerical positions. Racks are made of PP (filled PP, density 1.2 g/cm3), will not float in waterbath. Operating temperature: -20 to 90 °C. Autoclavable at 121 °C. Sturdy construction racks are supplied in two-pieces for convenient and permanent assembly. Available in four different colors.
Autres remarques
BR4341003 (India)
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG