Documents
T6316
Pharmed tubing
High grade for peristaltic pumps and cell culture, I.D. × O.D. 1/4 in. × 3/8 in.
Synonyme(s) :
Saint Gobain AY242017, Peristaltic Pump Tubing
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.35
Fabricant/nom de marque
Tygon AY242017
D.I. × D.E.
1/4 in. × 3/8 in.
Description générale
  • Outlasts silicone tubing in peristaltic pumps by up to 30 times
  • Can be autoclaved repeatedly
  • Heat weldable for sterile access
  • Documented biocompatibility to the ISO 109993 standard
  • Meets USP Class VI, FDA, and NSF criteria
Les clients ont également consulté
Slide 1 of 1