Principaux documents
RF45645P-EP
Rotor for Eppendorf® 5430/5430R Centrifuges
FA-45-64-5-PCR, non aerosol tight, to hold, 64 x 0.2 mL PCR tubes, lid
About This Item
Code UNSPSC :
41103909
Caractéristiques
lid
to hold 64 x 0.2 mL PCR tubes
non aerosol tight
strip rotor
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 022654209 (NA)
Eppendorf® 5427714006 (EU)
Paramètres
13,543 × g max. RCF
Technique(s)
PCR: suitable
centrifugation: suitable
Compatibilité
for use with centrifuge models 5430/5430R
Description générale
Rotors to fot Eppendorf centrifuge 5430 and 5430 R.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG