Principaux documents
OH30253008
Ohaus® Scout® Portable Touchscreen Balance
Model STX422, weighing capacity 420 g, resolution: 0.01 g, AC/DC input 110 V, US 3-pin plug (North America)
About This Item
Code UNSPSC :
41111502
Matériaux
metal weighing plate
Caractéristiques
CSA approved
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Ohaus® 30253008
Plage de mesure de la capacité de pesée
420 g, resolution: 0.01 g
Diam. de la nacelle de pesée
12 cm
Description générale
Ideal for laboratory and industrial applications, the OHAUS Scout features a user-friendly color touchscreen and a compact stackable design. Designed to deliver superior performance with rapid stability testing and high resolution weighing results, these portable scales set a new standard in weighing applications. Their features also include superior overload protection and advanced connectivity options.
Application
Weighing, piece counting,% weighing, check weighing, animal weighing / dynamic weighing, totalizing, density determination, display hold, mole weighing
Caractéristiques et avantages
Stability indicator, stackable design with storage cover included (except 1 mg models), mechanical and software overload / underload protection, low battery level indicator, automatic shutdown, auto-tare, slope calibration mass included with models up to 620g.
Informations légales
Ohaus is a registered trademark of Ohaus Corp.
Scout is a registered trademark of Ohaus Corp.