Principaux documents
HDP1096
Perfecta3D® hanging drop plate
size 96 wells , with lid and tray, polystyrene (untreated), sterile
Synonyme(s) :
3D, 3D Cell Culture
About This Item
Code UNSPSC :
41100000
Matériaux
polystyrene (untreated)
Stérilité
sterile
Conditionnement
pack of 8 ea individually packaged
Fabricant/nom de marque
3D Biomatrix HDP1096-8
Taille
96 wells , with lid and tray
Description générale
Perfecta3D® Hanging Drop Plates facilitate consistent and controllable formation of three-dimensional (3D) spheroids and cell aggregates.
Cells can be mixed prior to seeding in wells for homogeneous co-cultures, or added after spheroids are formed for patterned co-cultures. Stem cell aggregates like embryoid bodies can be cultured and studied without cellular contact with surfaces. Media can be changed easily.
One aggregate forms per well. The 96- and 384-well Perfecta3D® Hanging Drop Plates are compatible with automated liquid handling equipment. Drugs and assay reagents can be added directly to the wells. One spheroid per well and control over spheroid size means consistent data.
  • Easy to use
  • One spheroid or aggregate per well
  • Access cultures from the top of the plate: perform media changes, add compounds, or add cells for co-cultures
  • Compatible with liquid handling systems, microplate readers, and manual pipetting
Informations légales
Perfecta3D is a registered trademark of 3D Biomatrix
Michele Minopoli et al.
Cancers, 13(13) (2021-07-03)
Myxoid liposarcoma (MLPS) is the second most common subtype of liposarcoma and has tendency to metastasize to soft tissues. To date, the mechanisms of invasion and metastasis of MLPS remain unclear, and new therapeutic strategies that improve patients' outcomes are