Documents
F1911
Sigma-Aldrich
Nalgene® membrane forceps
stainless steel
Synonyme(s) :
tongs, tweezers
About This Item
Code UNSPSC :
41122405
eCl@ss :
32035504
Nomenclature NACRES :
NB.35
Matériaux
stainless steel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Nalgene DS0399-0001
Description générale
The unique cross-action design holds the membrane without applying pressure that may cause damage.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries