Key Documents
EP5804772008
Eppendorf® Adapter for fixed-angle-rotor 6 x 85 mL
holds 1 tube 15-18 mL, set of 2 pieces
About This Item
Code UNSPSC :
41103911
Nomenclature NACRES :
NB.82
Caractéristiques
holds 1 tube 15-18 mL
Conditionnement
set of 2 pieces
Fabricant/nom de marque
Eppendorf® 5804772008
Technique(s)
centrifugation: suitable
Compatibilité
for use with Rotor F-34-6-38
Informations légales
Eppendorf is a registered trademark of Eppendorf AG