Principaux documents
EP0030610326
Eppendorf® Lobind Microplate 384/V-PP, 384 wells
def. color (wells colorless), with barcode, polypropylene, pkg of 80 ea
About This Item
Code UNSPSC :
41122107
Matériaux
polypropylene
Conditionnement
pkg of 80 ea
Fabricant/nom de marque
Eppendorf® 0030610326
Technique(s)
PCR: suitable
Taille
384 wells
Description générale
Maximize your sample recovery with Eppendorf LoBind Plates. Eppendorf LoBind Plates are designed with a special two-component polymer mix that creates a hydrophilic surface to ensure optimal sample recovery rates. These plates are specifically designed for research or sensitive testing methods where they enable excellent results due to less sample binding to the plate surface.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG