Principaux documents
EP0030128664
Eppendorf® Twin tec PCR plates, 96 wells
green plate, conical bottom colorless wells, skirted, pkg of 25 ea
About This Item
Code UNSPSC :
41106300
Nomenclature NACRES :
NB.15
Matériaux
conical bottom colorless wells
green plate
polycarbonate
Stérilité
non-sterile
Caractéristiques
skirt (Skirted)
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Eppendorf® 0030128664
Technique(s)
PCR: suitable
Taille
96 wells
Volume maximum des puits
150 μm
Description générale
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG