Documents
EP0030128613
Eppendorf® Twin tec PCR plates, 96 wells
red plate, conical bottom colorless wells, semi-skirted, pkg of 25 ea
About This Item
Code UNSPSC :
41106300
Nomenclature NACRES :
NB.15
Matériaux
conical bottom colorless wells
polycarbonate
red plate
Stérilité
non-sterile
Caractéristiques
skirt (Semi skirted)
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Eppendorf® 0030128613
Technique(s)
PCR: suitable
Taille
96 wells
Volume maximum des puits
250 μm
Caractéristiques et avantages
  • Eppendorf® Twin tec PCR plate combine a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors.
  • Fits most thermal cyclers.
  • 250 μL max. well volume (when used with cap strips).
  • Improved well-to-well tolerance.
  • Ideal for quantitative real-time PCR as well – OptiTrack® matrix for faster sample identification and fewer pipetting errors.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG