Principaux documents
BAF367401025
Bel-Art® Earth Friendly Spoon
Wide Handle, capacity 25.0 mL
Synonyme(s) :
sampling spoon
About This Item
Code UNSPSC :
41122403
Nomenclature NACRES :
NB.35
Matériaux
(PLA (polylactic acid) resin )
natural
Conditionnement
package of 10
Fabricant/nom de marque
Bel-Art F36740-1025
Capacité
25.0 mL
Description générale
Lessen environmental impact with these PLA (polylactic acid) resin sampling spoons. The production of 100% plant-based PLA contributes less greenhouse gasses and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource. Disposal options include industrial composting for biodegradability, incineration, recycling and landfill. Use a separate spoon for each material sampled to avoid cross contamination.
Caractéristiques et avantages
Green alternative to petroleum-based sampling tools; helps meet sustainability goals
Comparable in strength and durability to polystyrene, polypropylene and other plastic resin sampling tools
Autres remarques
Wide handles let you grip firmly and provide just enough stiffness to easily sample packed powders and other firm materials
Unique flat bottom shape allows convenient duo use as weighing boats to save time and assure accuracy
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Les clients ont également consulté
Slide 1 of 1