Principaux documents
AXYPCR24B
Corning® Axygen® PCR Microplate
size 24 wells, conical bottom blue polypropylene wells, maximum volume 200 μL, pkg of 5x10plates/cs
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.22
Matériaux
conical bottom blue polypropylene wells
Stérilité
non-sterile
Fabricant/nom de marque
Axygen Inc PCR-24-B
Corning
Conditionnement
pkg of 5x10plates/cs
Volume maximum
200 μL
Taille
24 wells
Volume de travail
200 μL
Adéquation
suitable for PCR
Type de liaison
non-treated surface
Informations légales
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.