Principaux documents
GX308083068
Supelco
Semi Demountable Torch
for Agilent 4500/7500
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
SB.32
Qualité
for Agilent 4500/7500
Fabricant/nom de marque
(30-808-3068)
Compatibilité
Agilent/HP 4500
Agilent/HP 7500
Description générale
Includes:
  • 31-808-3051 (Quartz Torch Body for Agilent SDT)
  • 31-808-0773 (Tapered Quartz Injector 2.4mm)
  • 31-808-3067 (Torch Adaptor)
  • GAZ-0604 (GazFit Connectors, 2 for 6mm OD side arms, 2 for 4mm OD side arms (PKT.4))
  • 31-100-0106 (Extractor Tool for Semi Demountable Torch, 14.2mm (standard size))