Documents
1.05549
Supelco
TLC aluminum sheets, Silica gel 60 F₂₅₄
pkg of 20 sheets, sheet L × W 5 cm × 7.5 cm, aluminum support
About This Item
Code UNSPSC :
41115711
Nomenclature NACRES :
NB.21
Matériaux
aluminum support
silica gel 60 matrix
Niveau de qualité
Caractéristiques
binder Organic Polymer
fluorescent indicator
Conditionnement
pkg of 20 sheets
Technique(s)
thin layer chromatography (TLC): suitable
Épaisseur de la couche
200 μm
Feuille, L × l
5 cm × 7.5 cm
Taille des particules
10-12 μm
Dimension de pores
60 Å medium pore diameter
Température de stockage
2-30°C
Description générale
20 Aluminium sheets 5 x 7.5 cm
Our classical silica TLC plates are based on a combination of our proven silica gel 60 and the addition of a unique polymeric binder resulting in a very adherent and hard surface that will not crack or blister and even allow writing with a pensil on the surface without risk to damage the layer.
Application
Aluminum TLC plate, silica gel coated with flourescent indicator F254. The silica gel TLC plates come in a quantity of 20 size 5x7.5 cm for thin layer chromatography applications.
Liaison
Replaces: 5549-4; 5549
Remarque sur l'analyse
Specific surface area (according to BET; 5-Pt. measurement): 480 - 540 m²/g
Pore volume (N₂-isotherm): 0.74 - 0.84 ml/g
d 50 (laser diffraction, size distribution): 9.7 - 11.7 µm
Layer thickness: 165 - 235 µm
Deviation of layer thickness per plate: ≤ 40 µm
Chromatographic testing:
Colour test
hRf-values
- bleu vif organol, colour test, lipophile:
18 - 26
- Ceres Black G, colour test, lipophile: 40 - 50
- ceres Violet brn, colour test, lipophile: 59 - 69
Typical value determined an a conditioned sheet
Eluent: toluene ( 45% rel. humidity)