Z742587
Accuris™ M1 Grade Calibration Weight
size 2000 g, stainless steel
About This Item
Code UNSPSC :
41111500
Nomenclature NACRES :
NB.71
Produits recommandés
Matériaux
stainless steel
Conditionnement
pkg of 1
Fabricant/nom de marque
Benchmark Scientific W1005-2000
Taille
2000 g
Catégories apparentées
Description générale
- F1 Grade tolerance per OIML (International Organization of Legal Metrology)
- Works with all Accuris Lab balances
- Non-Magnetic Stainless Steel
- Includes a protective plastic case
Informations légales
Accuris is a trademark of Benchmark Scientific, Inc.