Documents
Z673277
Stuart melting point tubes
Sealed at one end
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.79
Matériaux
soda-lime glass tube
Caractéristiques
graduations
Fabricant/nom de marque
Stuart SMP10/1
Diamètre
1.3 mm
Longueur du tube
100 mm
Tube, D.E. × D.I. × épaisseur de la paroi
1.9 mm × 1.3 mm × 0.3 mm
Description générale
Made from soda glass, these tubes are easy to seal in a Bunsen flame and break into two. Supplied in robust tube holders.
Informations légales
Stuart is a trademark of Bibby Sterlin Ltd.