Z615668
Julabo® bath carrier trays for SW22/SW23
for assembling max 4 test tube racks
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.77
Produits recommandés
Fabricant/nom de marque
Julabo 8970369
Julabo
Catégories apparentées
Description générale
Basic stainless steel tray for assembling a maximum of 4 test tube racks: 8970380, 8970381, 8970382 or 8970383. For use with Julabo shaking water bath models SW22, SW23.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.