Principaux documents
Z615641
Julabo® bath carrier trays for SW22/SW23
to fit 5 Erlenmeyer flasks 1000 cc
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.77
Fabricant/nom de marque
Julabo 8970366
Julabo
Description générale
Erlenmeyer flasks can be positioned in the shaking water bath using the stainless steel tray. A maximum of 5 Erlenmeyer flasks with 1000 ml capacity fit on a standard tray.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.