Z552666
Aldrich® reverse-flow bubbler with SafetyBarb® hose connectors
capacity 25 mL
Synonyme(s) :
bubblers, scientific bubblers
About This Item
Produits recommandés
Capacité
25 mL
Catégories apparentées
Description générale
Monitors gas flow into, or evolution from a system. Capacity 25 mL per chamber. Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany