Z552461
Aldrich® glass thermometer well
fits flask, 12 L, immersion L 200 mm
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Produits recommandés
Joint
joint ST/NS24/40
Caractéristiques
fits flask 12 L
Description générale
Knobbed on sides for easy handling.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC