Z531006
Aldrich® Liebig condenser
Overall H 300 mm, joint: ST/NS 29/32
About This Item
Code UNSPSC :
41121800
Joint
joint: ST/NS29/32
H totale
300 mm
Description générale
Condensers have a standard taper female joint and a lower male drip tip-joint.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC