Principaux documents
Z408530
Scienceware® Vikem® vinyl-coated lead wire
wire diam. 5/16 in.
Synonyme(s) :
coil wire, wire spiral, wire vinyl
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.43
Matériaux
lead (coated)
Stérilité
non-sterile
Conditionnement
pack of 1
Fabricant/nom de marque
Bel-Art
Diam. du fil
5/16 in.
Poids
176 g
Description générale
Can be coiled into a helix, spiral, cone, or any irregular shape to fit around flasks and bottle necks to stabilize them in a water bath. 12 in. length. Coated with chemically-resistant red Vikem vinyl.
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vikem is a registered trademark of SP Scienceware, a division of SP