Principaux documents
Z305502
Sigma-Aldrich
Elite safety Goggles
About This Item
Code UNSPSC :
12352200
Fabricant/nom de marque
Bollé
Disponibilité
not available in USA and Canada
Description générale
No Fog PC lens fits over most prescription spectacles with exceptional panoramic vision and comfort to the wearer. This model is fitted with a foam band, but without ventilation holes to give protection against gases and vapours as well as dust. Fits over most prescription spectacles. BS2092.1CDG