Principaux documents
Z300861
Quickfit® Reduction Adapters
top female joint: ST/NS 14/23, bottom male joint: ST/NS 19/26
Synonyme(s) :
DA12
1 EA
28.40 CHF
28.40 CHF
Date d'expédition estimée le22 mars 2025
Changer de vue
1 EA
28.40 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
28.40 CHF
Date d'expédition estimée le22 mars 2025
Joint
female joint: ST/NS top14/23
male joint: ST/NS bottom19/26
Fabricant/nom de marque
Quickfit DA 12
Disponibilité
not available in USA and Canada
Description générale
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Informations légales
Quickfit is a registered trademark of SciLabware Limited