Documents
Z285498
Sigma-Aldrich
DigiTrol II temperature controller
AC/DC input 120 V AC, 15 A
Synonyme(s) :
Temperature controller, Temperature monitoring system
About This Item
Code UNSPSC :
41112303
Nomenclature NACRES :
NB.76
Fabricant/nom de marque
Glas-Col 104APL612K
Disponibilité
not available in EU
Paramètres
32-999 °F temperature (0-750 °C)
Sortie
1,800 watts
Description générale
Simultaneously displays set point and bath temperature to 1.0°C. Supplied with a Type K, 6 in. immersion thermocouple probe that fits Aldrich oil baths. 240 V models are supplied with a US connector.