Z270385
Support ring
ring diam. 2 in.
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
Diam. de l'anneau
2 in.
Description générale
Cast iron ring with integral support clamp to attach to standard supports. Fits 1/2 in. diameter rods.