Z117447
LDPE laboratory tubing
I.D. × O.D. 3/16 in. × 5/16 in.
About This Item
Code UNSPSC :
41100000
Produits recommandés
D.I. × D.E.
3/16 in. × 5/16 in.
Épaisseur de la paroi
1/16 in.
Description générale
Good chemical resistance, translucent, and rigid. Temp. range: -103 to 120°F.