Principaux documents
Z104876
PTFE sleeve
joint: ST/NS 14/35
Synonyme(s) :
ptfe sleeves
About This Item
Code UNSPSC :
41100000
Joint
joint: ST/NS14/35
Conditionnement
pkg of 6 ea
Fabricant/nom de marque
Bel-Art
Description générale
Extremely thin, accurately tapered, slip-on tubes for ground-glass joints prevent seizing and galling. Chemically inert and used without lubricants or adhesives, thus precluding contamination. Temperature range: −100 to 250°C.