Principaux documents
TEM-LC200NI25
Sigma-Aldrich
Lacey Carbon Supported Nickel Grids
size 200 mesh, size 50 nm , hole size, box of 25 ×
Synonyme(s) :
Lacey/Carbon, 200 Mesh, Nickel, 25pk
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 25 ×
Taille
200 mesh
50 nm , hole size
Description générale
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.