TEM-CF400NI
Carbon Film Supported Nickel Square Mesh
size 400 mesh, box of 25
Synonyme(s) :
CARBON FILM 400# NICKEL 25/PK
About This Item
Code UNSPSC :
41111761
Conditionnement
box of 25
Taille
400 mesh
Description générale
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Nickel grid, 400 mesh.