Documents
TEM-CF200NI50
Sigma-Aldrich
Carbon Film Supported Nickel Square Mesh
size 200 mesh, box of 50
Synonyme(s) :
CARBON FILM 200 MESH NI 50/BX
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 50
Taille
200 mesh
Description générale
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Nickel grid, 200 mesh.