Principaux documents
SYNA831922
Synthware inlet adapter with fritted disc
joint: ST/NS 19/22
Synonyme(s) :
Synthware A831922, glass inlet adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A831922
Description générale
With an extra coarse disc to prevent solid from being introduced into reaction system during liquid transfer.
Informations légales
Synthware is a trademark of Kemtech America Inc.