Documents
SYNA621420
Synthware vacuum take-off adapter with short stem
joint: ST/NS 14/20, hose size 8 mm
Synonyme(s) :
Synthware A621420, distillation adapter, vacuum take off adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A621420
Taille du tuyau
8 mm
Description générale
With a hose connection and a drip tip.
Informations légales
Synthware is a trademark of Kemtech America Inc.