Principaux documents
DWK07-3000
Wheaton® Microliter μlplates Deep Polypropylene
About This Item
Code UNSPSC :
41106200
Nomenclature NACRES :
NB.15
Matériaux
polypropylene plate
round bottom
round wells
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
WHEATON® 07-3000
Capacité
1.3 mL
Application
Chromatography
Caractéristiques et avantages
These products are sold as one microplate with 96 inserts installed.
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.