Principaux documents
CLS88202429
Pyrex® enlarging adapter
female joint: ST/NS 24/40, male joint: ST/NS 29/42
Synonyme(s) :
lab equipment accessory
About This Item
Code UNSPSC :
41121815
Joint
female joint: ST/NS24/40
male joint: ST/NS29/42
Fabricant/nom de marque
Corning 8820-2429
Disponibilité
not available in EU
Longueur
85 mm
Informations légales
PYREX is a registered trademark of Corning, Inc.