Principaux documents
CLS70360125
Pyrex® VISTA filter flask
capacity 125 mL
Synonyme(s) :
conical flasks, filtering flask, filtering flasks, flasks erlenmeyer, Corning® 70360
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
flat bottom flask
narrow-mouth
Stérilité
non-sterile
Caractéristiques
closure type ground joint or rubber stopper
Conditionnement
case of 12
pack of 6
Fabricant/nom de marque
Corning 70360-125
Capacité
125 mL
Catégories apparentées
Description générale
These small filtering flasks are recommended for microchemical use. All flasks have permanent blue enamel marking spots.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.