Documents
BR782891
BRAND® BRANDplates® for inserts
6-well with lid cellGrade plus
Synonyme(s) :
BRANDplates®, Cell culture, Insert System, Inserts, Plates, Tissue culture
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
transparent wells
Stérilité
sterile
Caractéristiques
lid
single packed
Conditionnement
pack of 1 ea
pkg of Individually wrapped, with lid
Fabricant/nom de marque
BRAND 782891
Taille
6 well
Description générale
PS. cellGrade plus surface, sterile, hydrophilic with different chemical groups, such as carboxyl and hydroxyl groups, that are freely accessible.Cover with condensation rings.Manufactured according to the requirements in ANSI/SLAS Standards 1 and 4.BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic
Caractéristiques et avantages
• Optimal cell growth thanks to cellGrade plus surface• Side well access for easier pipetting and removing cover slips• Perfect positioning of the BRANDplates® insert
Informations légales
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG