Principaux documents
BR130426
BRAND® square bottle with tamper-proof screw cap, PE-HD
capacity 2500 mL, wide-neck
Synonyme(s) :
Bottles, Plastics, Square bottles
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux
high-density polyethylene bottle
polypropylene cap
square bottle
translucent bottle
wide-mouth bottle
wide-neck
Joint
Joints threaded neck (GL80)
Caractéristiques
with cap
Conditionnement
pack of 100 ea
Fabricant/nom de marque
BRAND 130426
Long. × Haut.
112 mm × 258 mm
Capacité
2500 mL
Description générale
Translucent. Screw cap with sealing cone, PP, blue, with cadmium-free pigments. Tamper-proof closure with press-off retaining ring. Space-saving square base.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG