Merck
  • Microfluidic conceived Trojan microcarriers for oral delivery of nanoparticles.
Microfluidic conceived Trojan microcarriers for oral delivery of nanoparticles.
International journal of pharmaceutics (2015-06-28)
Ikram Ullah Khan, Christophe A Serra, Nicolas Anton, Mériem Er-Rafik, C Blanck, Marc Schmutz, Isabelle Kraus, Nadia Messaddeq, Christophe Sutter, Halina Anton, Andrey S Klymchenko, Thierry F Vandamme
ABSTRACT
In this study, we report on a novel method for the synthesis of poly(acrylamide) Trojan microparticles containing ketoprofen loaded poly(ethyl acrylate) or poly(methyl acrylate) nanoparticles. To develop these composite particles, a polymerizable nanoemulsion was used as a template. This nanoemulsion was obtained in an elongational-flow micromixer (μRMX) which was linked to a capillary-based microfluidic device for its emulsification into micron range droplets. Downstream, the microdroplets were hardened into Trojan particles in the size range of 213-308 μm by UV initiated free radical polymerization. The nanoemulsion size varied from 98 -132 nm upon changes in surfactant concentration and number of operating cycles in μRMX. SEM and confocal microscopy confirmed the Trojan morphology. Under SEM it was observed that the polymerization reduced the size of the nanoemulsion down to 20-32 nm for poly(ethyl acrylate) and 10-15 nm for poly(methyl acrylate) nanoparticles. This shrinkage was confirmed by cryo-TEM studies. We further showed that Trojan microparticles released embedded nanoparticles on contact with suitable media as confirmed by transmission electron microscopy. In a USP phosphate buffer solution of pH 6.8, Trojan microparticles containing poly(ethyl acrylate) nanoparticles released 35% of encapsulated ketoprofen over 24h. The low release of the drug was attributed to the overall low concentration of nanoparticles and attachment of some of nanoparticles to the poly(acrylamide) matrix. Thus, this novel method has shown possibility to develop Trojan particles convieniently with potential to deliver nanoparticles in the gastrointestinal tract.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
1-Hydroxycyclohexyl phenyl ketone, 99%
Sigma-Aldrich
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Sigma-Aldrich
Methyl acrylate, 99%, contains ≤100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, for molecular biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Ethyl acrylate, ≥99.5%, stabilized
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%