Merck
  • Tyrosine kinase inhibitor tyrphostin AG490 triggers both apoptosis and autophagy by reducing HSF1 and Mcl-1 in PEL cells.
Tyrosine kinase inhibitor tyrphostin AG490 triggers both apoptosis and autophagy by reducing HSF1 and Mcl-1 in PEL cells.
Cancer letters (2015-07-18)
Marisa Granato, Barbara Chiozzi, Maria Rosaria Filardi, Lavinia Vittoria Lotti, Livia Di Renzo, Alberto Faggioni, Mara Cirone
ABSTRACT
PEL cells relay on the constitutive activation of STAT3 for their survival, thus its inhibition by AG490 leads to apoptotic cell death. In this study, we found that the cytotoxic activity of AG490 correlated with the reduction of HSP70 and its master regulator HSF1 that, based on knocking-down experiments, was found to play a pro-survival role in PEL cells. To counteract the pro-death effect mediated by HSF1/HSP70 down-regulation, AG490 induced a complete autophagy, whose inhibition potentiated its cytotoxic effect against PEL cells. AG490 as well as HSF1 siRNA reduced the expression of Mcl-1, a Bcl-2 family member that negatively regulates apoptosis and autophagy. These results suggest that STAT3 inhibition, by down-regulating the expression of HSF1/HSP70, reduces Mcl-1 and leads to both apoptosis and autophagy induction in PEL cells.
Product Description
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Atp11c
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
SAFC
BIS-TRIS
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)