Merck
  • Emilin1 deficiency causes structural and functional defects of lymphatic vasculature.
Emilin1 deficiency causes structural and functional defects of lymphatic vasculature.
Molecular and cellular biology (2008-04-16)
Carla Danussi, Paola Spessotto, Alessandra Petrucco, Bruna Wassermann, Patrizia Sabatelli, Monica Montesi, Roberto Doliana, Giorgio M Bressan, Alfonso Colombatti
ABSTRACT
Lymphatic-vasculature function critically depends on extracellular matrix (ECM) and on its connections with lymphatic endothelial cells (LECs). However, the composition and the architecture of ECM have not been fully taken into consideration in studying the biology and the pathology of the lymphatic system. EMILIN1, an elastic microfibril-associated protein, is highly expressed by LECs in vitro and colocalizes with lymphatic vessels in several mouse tissues. A comparative study between WT and Emilin1-/- mice highlighted the fact that Emilin1 deficiency in both CD1 and C57BL/6 backgrounds results in hyperplasia, enlargement, and frequently an irregular pattern of superficial and visceral lymphatic vessels and in a significant reduction of anchoring filaments. Emilin1-deficient mice also develop larger lymphangiomas than WT mice. Lymphatic vascular morphological alterations are accompanied by functional defects, such as mild lymphedema, a highly significant drop in lymph drainage, and enhanced lymph leakage. Our findings demonstrate that EMILIN1 is involved in the regulation of the growth and in the maintenance of the integrity of lymphatic vessels, a fundamental requirement for efficient function. The phenotype displayed by Emilin1(-/-) mice is the first abnormal lymphatic phenotype associated with the deficiency of an ECM protein and identifies EMILIN1 as a novel local regulator of lymphangiogenesis.
Sigma-Aldrich
Evans Blue, Dye content ≥75 %
Sigma-Aldrich
Freund′s Adjuvant, Incomplete, liquid
Sigma-Aldrich
Mineral oil, heavy
Sigma-Aldrich
Mineral oil, light
Sigma-Aldrich
Mineral oil, light oil, suitable for mouse embryo cell culture, BioXtra
Sigma-Aldrich
Mineral oil, BioUltra, for molecular biology
Sigma-Aldrich
Mineral oil, BioReagent, for molecular biology, light oil
Sigma-Aldrich
Mineral oil, meets USP testing specifications
Sigma-Aldrich
Mineral oil, suitable for preparation of Nujol mulls for infrared spectroscopy, light oil
Sigma-Aldrich
Mineral oil, PCR Reagent
Sigma-Aldrich
Mineral oil, light oil, (neat)