Merck
  • A high-frequency regulatory polymorphism in the p53 pathway accelerates tumor development.
A high-frequency regulatory polymorphism in the p53 pathway accelerates tumor development.
Cancer cell (2010-09-14)
Sean M Post, Alfonso Quintás-Cardama, Vinod Pant, Tomoo Iwakuma, Amir Hamir, James G Jackson, Daniela R Maccio, Gareth L Bond, David G Johnson, Arnold J Levine, Guillermina Lozano
ABSTRACT
MDM2, a negative regulator of p53, is elevated in many cancers that retain wild-type p53. A single nucleotide polymorphism (SNP) in the human MDM2 promoter increases the affinity of Sp1 resulting in elevated MDM2 levels. We generated mice carrying either the MDM2(SNP309T) or the MDM2(SNP309G) allele to address the impact of MDM2(SNP309G) on tumorigenesis. Mdm2(SNP309G/G) cells exhibit elevated Mdm2 levels, reduced p53 levels, and decreased apoptosis. Importantly, some Mdm2(SNP309G/G) mice succumbed to tumors before 1 year of age, suggesting that this allele increases tumor risk. Additionally, the Mdm2(SNP309G) allele potentiates the tumor phenotype and alters tumor spectrum in mice inheriting a p53 hot-spot mutation. These data provide causal evidence for increased cancer risk in carriers of the Mdm2(SNP309G) allele.
Sigma-Aldrich
Anti-MDM2 Antibody, clone 2A10, clone 2A10, from mouse