Merck
  • Balancing of histone H3K4 methylation states by the Kdm5c/SMCX histone demethylase modulates promoter and enhancer function.
Balancing of histone H3K4 methylation states by the Kdm5c/SMCX histone demethylase modulates promoter and enhancer function.
Cell reports (2013-04-03)
Nikolay S Outchkourov, Jose M Muiño, Kerstin Kaufmann, Wilfred F J van Ijcken, Marian J Groot Koerkamp, Dik van Leenen, Petra de Graaf, Frank C P Holstege, Frank G Grosveld, H T Marc Timmers
ABSTRACT
The functional organization of eukaryotic genomes correlates with specific patterns of histone methylations. Regulatory regions in genomes such as enhancers and promoters differ in their extent of methylation of histone H3 at lysine-4 (H3K4), but it is largely unknown how the different methylation states are specified and controlled. Here, we show that the Kdm5c/Jarid1c/SMCX member of the Kdm5 family of H3K4 demethylases can be recruited to both enhancer and promoter elements in mouse embryonic stem cells and in neuronal progenitor cells. Knockdown of Kdm5c deregulates transcription via local increases in H3K4me3. Our data indicate that by restricting H3K4me3 modification at core promoters, Kdm5c dampens transcription, but at enhancers Kdm5c stimulates their activity. Remarkably, an impaired enhancer function activates the intrinsic promoter activity of Kdm5c-bound distal elements. Our results demonstrate that the Kdm5c demethylase plays a crucial and dynamic role in the functional discrimination between enhancers and core promoters.
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, clone 15-10C-E4, rabbit monoclonal, clone 15-10C-E4, Upstate®, from rabbit
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)