Skip to Content
Merck
All Photos(1)

Key Documents

V900762

Sigma-Aldrich

9,10-Phenanthrenequinone

Vetec, reagent grade, 94%

Synonym(s):

9,10-Phenanthrenedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O2
CAS Number:
Molecular Weight:
208.21
Beilstein:
608838
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

94%

mp

209-212 °C (lit.)

SMILES string

O=C1C(=O)c2ccccc2-c3ccccc13

InChI

1S/C14H8O2/c15-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14(13)16/h1-8H

InChI key

YYVYAPXYZVYDHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

465.8 °F - closed cup

Flash Point(C)

241 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiyuki Matsunaga et al.
Free radical research, 45(7), 848-857 (2011-06-01)
9,10-Phenanthrenequinone (9,10-PQ), a redox-active quinone in diesel exhausts, triggers cellular apoptosis via reactive oxygen species (ROS) generation in its redox cycling. This study found that induction of CCAAT/enhancer-binding protein-homologous protein (CHOP), a pro-apoptotic factor derived from endoplasmic reticulum stress, participates
Pedro Sanchez-Cruz et al.
Chemical research in toxicology, 22(5), 818-823 (2009-03-24)
The quinones 1,4-naphthoquinone, methyl-1,4-naphthoquinone, tetramethyl-1,4-benzoquinone, 2,3-dimethoxy-5-methyl-1,4-benzoquinone, 2,6-dimethylbenzoquinone, 2,6-dimethoxybenzoquinone, and 9,10-phenanthraquinone enhance the rate of nitric oxide reduction by xanthine/xanthine oxidase in nitrogen-saturated phosphate buffer (pH 7.4). Maximum initial rates of NO reduction (V(max)) and the amount of nitrous oxide produced
Toshiyuki Matsunaga et al.
Cell and tissue research, 347(2), 407-417 (2012-01-28)
9,10-Phenanthrenequinone (9,10-PQ), a major quinone in diesel exhaust particles, induces apoptosis via the generation of reactive oxygen species (ROS) because of 9,10-PQ redox cycling. We have found that intratracheal infusion of 9,10-PQ facilitates the secretion of surfactant into rat alveolus.
Petr Milko et al.
Inorganic chemistry, 48(24), 11734-11742 (2009-11-26)
With the use of the model complexes [(PQ)FeCl(CH(3)O)](+), [(phen)FeCl(CH(3)O)](+), and [(PQ)(phen)FeCl(CH(3)O)](+), where PQ is 9,10-phenanthraquinone and phen is 1,10-phenanthroline, the reactivity of phenanthraquinone in complexes with iron(III) is investigated. It is shown that 9,10-phenanthraquinone takes part in redox processes occurring
Keiko Taguchi et al.
Free radical biology & medicine, 44(8), 1645-1655 (2008-02-26)
9,10-Phenanthraquinone (PQ), a component of airborne particulate matter, causes marked cellular protein oxidation and cytotoxicity through a two-electron reduction to 9,10-dihydroxyphenanthrene (PQH2), which is associated with the propagation of reactive oxygen species (K. Taguchi et al., Free Radic. Biol. Med.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service