Skip to Content
Merck
All Photos(1)

Documents

V900723

Sigma-Aldrich

5-Methylfurfural

Vetec, reagent grade, 98%

Synonym(s):

5-Methyl-2-furaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O2
CAS Number:
Molecular Weight:
110.11
Beilstein:
106895
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

refractive index

n20/D 1.531 (lit.)

bp

187 °C (lit.)

density

1.107 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1ccc(C)o1

InChI

1S/C6H6O2/c1-5-2-3-6(4-7)8-5/h2-4H,1H3

InChI key

OUDFNZMQXZILJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Pitié et al.
Nucleic acids research, 28(24), 4856-4864 (2000-01-11)
Mechanisms of DNA oxidation by copper complexes of 3-Clip-Phen and its conjugate with a distamycin analogue, in the presence of a reductant and air, were studied. Characterisation of the production of 5-methylenefuranone (5-MF) and furfural, associated with the release of
E M Jouad et al.
Journal of inorganic biochemistry, 86(2-3), 565-571 (2001-09-22)
5-Methyl 2-furfuraldehyde thiosemicarbazone (M5HFTSC) with nickel(II) leads to three types of complexes: [Ni(M5HFTSC)(2)X(2)], [Ni(M5FTSC)(2)] and [Ni(M5FTSC)(2)] x 2DMF. In the first type the ligand remains in thione form, while in the two other, the anionic thiolato form is involved. The
S Uddin et al.
Biochemistry and molecular biology international, 35(1), 185-195 (1995-01-01)
Furfural and methylfurfural are dietary mutagens and are present in various food products and beverages. The alkaline-induced unwinding of calf thymus DNA permitted the measurement of the number of strand breaks formed by furfural and methylfurfural, as a function of
P Rao et al.
Analytical biochemistry, 189(2), 178-181 (1990-09-01)
Even though the chromogens formed from mannose and galactose showed comparable absorbances at 480 nm in the conventional (developer present during heat of dilution) and modified (developer reacted at room temperature after cooling; epsilon mannose = 13,700, galactose = 14,000)
[Gas-chromatographic determination of furfural, methylfurfural and furyl alcohol in air].
V V Tarasov
Gigiena truda i professional'nye zabolevaniia, (12)(12), 53-54 (1985-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service