Skip to Content
Merck
All Photos(3)

Key Documents

V900675

Sigma-Aldrich

Tropic acid

Vetec, reagent grade, 98%

Synonym(s):

2-Phenylhydracrylic acid, 3-Hydroxy-2-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH2OH)CO2H
CAS Number:
Molecular Weight:
166.17
Beilstein:
2209199
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

98%

mp

116-118 °C (lit.)

SMILES string

OCC(C(O)=O)c1ccccc1

InChI

1S/C9H10O3/c10-6-8(9(11)12)7-4-2-1-3-5-7/h1-5,8,10H,6H2,(H,11,12)

InChI key

JACRWUWPXAESPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick K Harrison et al.
Naunyn-Schmiedeberg's archives of pharmacology, 373(3), 230-236 (2006-06-01)
The enzyme atropinesterase (EC 3.1.1.10) causes the rapid hydrolysis of tropane alkaloids such as atropine and scopolamine. This enzyme is known to occur in a certain proportion of rabbits and some plants, although its presence in other animal species remains
The biosynthesis of shikimate metabolites.
A R Knaggs
Natural product reports, 18(3), 334-355 (2001-07-31)
Stephanie Heine et al.
Electrophoresis, 24(15), 2687-2692 (2003-08-06)
A method for the separation of atropine enantiomers, D- and L-hyoscyamine by capillary electrophoresis (CE) has been developed and validated. The advantages of the CE method compared with polarimetry include smaller amounts of analytes and a lower limit of detection
Simultaneous determination of atropine sulphate and tropic acid by reversed phase high-pressure liquid chromatography.
A Richard et al.
Die Pharmazie, 39(12), 866-867 (1984-12-01)
M Eeva et al.
Journal of pharmaceutical and biomedical analysis, 16(5), 717-722 (1998-04-16)
A capillary zone electrophoretic method (CZE) was developed using an uncoated fused silica capillary for the separation and determination of the main tropane alkaloids. The applicability of the developed method for analysis of plant samples was examined by analyzing samples

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service