Skip to Content
Merck
All Photos(2)

Documents

V900603

Sigma-Aldrich

3,4-Dihydroxybenzaldehyde

Vetec, reagent grade, 97%

Synonym(s):

Protocatechualdehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CHO
CAS Number:
Molecular Weight:
138.12
Beilstein:
774381
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

97%

mp

150-157 °C (lit.)

SMILES string

Oc1ccc(C=O)cc1O

InChI

1S/C7H6O3/c8-4-5-1-2-6(9)7(10)3-5/h1-4,9-10H

InChI key

IBGBGRVKPALMCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun-Jing Zhang et al.
Journal of ethnopharmacology, 136(1), 129-136 (2011-04-30)
To evaluate the pharmacokinetics of phenolic compounds after oral administration of Danshen extract in rat brain. Blood and brain microdialysis probes were inserted into jugular vein and cerebral cortex of rat under anesthesia and perfused with ringer's solution at the
Yong Xu et al.
Basic & clinical pharmacology & toxicology, 110(4), 384-389 (2011-11-05)
Recent studies have demonstrated that nuclear factor-κB (NF-κB) and high-mobility group box 1 (HMGB1) are associated with the pathophysiology of sepsis. The present study was carried out to investigate the effects of protocatechuic aldehyde (PA) on an experimental model of
Zhi-Qiang Chang et al.
Journal of nutritional science and vitaminology, 57(1), 118-122 (2011-04-23)
Although various biological activities of Phellinus gilvus (PG) have been reported, the active compounds responsible for these effects are not known. Here, we evaluated the activity of various solvent extracts of PG, and found the ethyl acetate extract (Fd) to
Ya-Ling Xing et al.
Phytotherapy research : PTR, 26(9), 1334-1341 (2012-02-03)
Apoptosis of vascular endothelial cells results in the loss of endothelial integrity, and is a risk factor of atherosclerosis (AS). Lipopolysaccharide (LPS) stimulates inflammation during AS. The current study examined the effect of a potent water-soluble antioxidant, protocatechuic aldehyde (PCA;
Yuzo Nakagawa-Yagi et al.
BMC complementary and alternative medicine, 12, 101-101 (2012-07-20)
Extracts of Sasa senanensis Rehder are used in traditional Japanese medicine; however, little is known about the underlying mechanisms of their potential health benefits. S. senanensis leaves were extracted with subcritical water. An active small-molecule was isolated using reversed-phase high-performance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service