Skip to Content
Merck
All Photos(1)

Key Documents

V900577

Sigma-Aldrich

3,4,5-Trimethoxybenzoic acid

Vetec, reagent grade, 99%

Synonym(s):

Gallic acid trimethyl ether, Trimethylgallic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CO2H
CAS Number:
Molecular Weight:
212.20
Beilstein:
884655
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

bp

225-227 °C/10 mmHg (lit.)

mp

168-171 °C (lit.)

SMILES string

COc1cc(cc(OC)c1OC)C(O)=O

InChI

1S/C10H12O5/c1-13-7-4-6(10(11)12)5-8(14-2)9(7)15-3/h4-5H,1-3H3,(H,11,12)

InChI key

SJSOFNCYXJUNBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Naviasky
Journal of pharmaceutical sciences, 73(4), 542-545 (1984-04-01)
An ion-pair column chromatographic/UV spectrophotometric method for assaying trimethobenzamide hydrochloride in capsules and injections is presented, as well as a method for the detection of 3,4,5- trimethoxybenzoic acid in trimethobenzamide hydrochloride bulk drug and dosage forms. Results obtained by the
E N Schachter et al.
Respiration; international review of thoracic diseases, 65(5), 393-400 (1998-10-23)
Latex manufacturing workers are exposed to a heterogeneous aerosol of organic compounds. Previous studies of latex workers involved in glove production indicate that these individuals are at risk of developing respiratory symptoms and impaired lung function. The effect of latex
H Tateno et al.
Mutation research, 327(1-2), 237-246 (1995-03-01)
Many inhibitors of tubulin polymerization have a trimethoxybenzene ring in their molecules. Such trimethoxybenzoic compounds and their analogues may therefore have a potency to induce meiotic nondisjunction of oocytes. In this study, a single dose of reserpine (0.5 microgram/g body
S Kleis-San Francisco et al.
The Journal of experimental zoology, 244(1), 133-143 (1987-10-01)
In the amphibian ovarian follicle, progesterone production is thought to induce maturation of the enclosed oocyte. Intracellular mechanisms regulating these events in the somatic and germ cells are incompletely understood. However, calcium appears to play a role in the production
[Synthesis and the study of the effect of new 3,4,5-trimethoxybenzoic acid derivatives on the central nervous system].
R Glinka et al.
Acta poloniae pharmaceutica, 42(2), 117-122 (1985-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service