Skip to Content
Merck
All Photos(3)

Key Documents

V900479

Sigma-Aldrich

HEPES sodium salt

Vetec, reagent grade, 96%

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17N2NaO4S
CAS Number:
Molecular Weight:
260.29
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

96%

form

powder

useful pH range

6.8-8.2

pKa (25 °C)

7.5

solubility

water: 500 mg/mL, clear, colorless

SMILES string

[Na+].OCCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O4S.Na/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14;/h11H,1-8H2,(H,12,13,14);/q;+1/p-1

InChI key

RDZTWEVXRGYCFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Yih Jeng et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 7), 829-838 (2012-07-04)
NkBgl, a β-glucosidase from Neotermes koshunensis, is a β-retaining glycosyl hydrolase family 1 enzyme that cleaves β-glucosidic linkages in disaccharide or glucose-substituted molecules. β-Glucosidases have been widely used in several applications. For example, mutagenesis of the attacking nucleophile in β-glucosidase
Junbo Li et al.
Dalton transactions (Cambridge, England : 2003), 41(13), 3623-3626 (2012-02-24)
We developed a novel method for Cu(2+) detection based on the ion promoted oxidation reaction. Chemodosimeter L (weak fluorescence) can be oxidized into 3-benzothiazoly-7-N,N-diethylaminocoumarin (strong green fluorescence, coumarin 6) by Cu(2+) with high selectivity and sensitivity in HEPES (10 mM
Rakesh K Pathak et al.
Analytical chemistry, 84(11), 5117-5123 (2012-05-04)
An in situ prepared Zn(2+) complex of triazole linked imino-thiophenyl conjugate of calix[4]arene, [ZnL], was demonstrated to be highly fluorescent in HEPES buffer solution. [ZnL] has been used as a chemo-sensing ensemble for the recognition of phosphates in general and
Jemimah Walker et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 100(4), 1134-1141 (2012-02-15)
Magnesium (Mg) and its alloys have been proposed as degradable replacements to commonly used orthopedic biomaterials such as titanium alloys and stainless steel. However, the corrosion of Mg in a physiological environment remains a difficult characteristic to accurately assess with
Anca G Delgado et al.
Microbial cell factories, 11, 128-128 (2012-09-15)
Buffering to achieve pH control is crucial for successful trichloroethene (TCE) anaerobic bioremediation. Bicarbonate (HCO3-) is the natural buffer in groundwater and the buffer of choice in the laboratory and at contaminated sites undergoing biological treatment with organohalide respiring microorganisms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service